Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI2 SIRNA Assays ›
Gene Symbol:
ABI2
Gene Name:
abl-interactor 2
Gene Aliases:
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
Chromosome Location:
Chr.2: 203327874 - 203432173 on Build GRCh38
UniGene ID:
Hs.471156
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148366
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282925.1
NP_001269854.1
2
530
NM_001282926.1
NP_001269855.1
2
530
NM_001282927.1
NP_001269856.1
1
130
NM_001282932.1
NP_001269861.1
2
526
NM_005759.5
NP_005750.4
2
530
XM_005246217.2
XP_005246274.1
2
895
XM_005246218.2
XP_005246275.1
2
895
XM_005246221.2
XP_005246278.1
2
895
XM_005246225.2
XP_005246282.1
2
895
XM_005246226.2
XP_005246283.1
2
895
XM_006712174.2
XP_006712237.1
2
895
XM_006712177.2
XP_006712240.1
2
895
XM_006712179.2
XP_006712242.1
2
895
XM_006712180.2
XP_006712243.1
2
895
XM_006712181.2
XP_006712244.1
2
895
XM_006712182.2
XP_006712245.1
2
895
XM_006712185.1
XP_006712248.1
2
895
XM_006712186.2
XP_006712249.1
2
895
XM_011510472.2
XP_011508774.1
2
895
XM_011510473.2
XP_011508775.1
1
137
XM_017003118.1
XP_016858607.1
2
895
XM_017003119.1
XP_016858608.1
2
895
XM_017003120.1
XP_016858609.1
2
895
XM_017003121.1
XP_016858610.1
2
895
XM_017003122.1
XP_016858611.1
2
895
XM_017003123.1
XP_016858612.1
2
895
XM_017003124.1
XP_016858613.1
2
895
XM_017003125.1
XP_016858614.1
2
895
XM_017003126.1
XP_016858615.1
2
895
XM_017003127.1
XP_016858616.1
2
895
XM_017003128.1
XP_016858617.1
2
895
XM_017003129.1
XP_016858618.1
2
895
XM_017003130.1
XP_016858619.1
2
895
XM_017003131.1
XP_016858620.1
2
895
XM_017003132.1
XP_016858621.1
2
895
XM_017003133.1
XP_016858622.1
2
895
XM_017003134.1
XP_016858623.1
2
895
XM_017003135.1
XP_016858624.1
2
895
XM_017003136.1
XP_016858625.1
2
895
XM_017003137.1
XP_016858626.1
2
895
XM_017003138.1
XP_016858627.1
2
895
XM_017003139.1
XP_016858628.1
2
895
XM_017003140.1
XP_016858629.1
2
895
XM_017003141.1
XP_016858630.1
2
895
XM_017003142.1
XP_016858631.1
2
895
XM_017003143.1
XP_016858632.1
2
895
XM_017003144.1
XP_016858633.1
2
895
XM_017003145.1
XP_016858634.1
2
895
XM_017003146.1
XP_016858635.1
2
895
GenBank mRNA
AF260261.1
-
2
254
AK299824.1
-
2
374
AK302853.1
-
2
452
AK316000.1
-
2
360
BC001439.2
-
2
279
3138341
BT009920.1
-
2
254
DC376086.1
-
2
379
U23435.1
-
1
91
X95632.1
-
2
354
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
cytoskeleton organization
actin polymerization or depolymerization
viral process
cell migration
Rac protein signal transduction
peptidyl-tyrosine phosphorylation
positive regulation of Arp2/3 complex-mediated actin nucleation
DNA binding
protein binding
cytoskeletal adaptor activity
SH3 domain binding
kinase binding
ubiquitin protein ligase binding
protein complex binding
Rac GTPase binding
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0