Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RHOT1 SIRNA Assays ›
Gene Symbol:
RHOT1
Gene Name:
ras homolog family member T1
Gene Aliases:
ARHT1, MIRO-1, MIRO1
Chromosome Location:
Chr.17: 32142454 - 32253374 on Build GRCh38
UniGene ID:
Hs.655325
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148717
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RHOT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033566.2
NP_001028738.1
8
718
NM_001033567.2
NP_001028739.2
9
842
NM_001033568.2
NP_001028740.1
8
718
NM_001288754.1
NP_001275683.1
8
718
NM_001288755.1
NP_001275684.1
8
714
NM_001288758.1
NP_001275687.1
9
842
NM_018307.4
NP_060777.3
8
718
NR_110083.1
8
718
XM_011524969.2
XP_011523271.1
7
442
XM_011524971.2
XP_011523273.2
8
514
XM_011524973.2
XP_011523275.2
8
514
XM_011524975.2
XP_011523277.1
9
971
XM_011524976.2
XP_011523278.2
8
514
XM_017024819.1
XP_016880308.1
9
746
XM_017024820.1
XP_016880309.1
7
442
XM_017024821.1
XP_016880310.1
8
514
XM_017024822.1
XP_016880311.1
8
743
XM_017024823.1
XP_016880312.1
8
514
XM_017024824.1
XP_016880313.1
9
849
XM_017024825.1
XP_016880314.1
9
941
GenBank mRNA
AJ496730.1
-
8
706
AJ517412.1
-
8
479
AK001902.1
-
2
82
AK022695.1
-
1
12
AK294407.1
-
8
492
AL136929.1
-
8
542
AY094972.1
-
8
595
BC015698.2
-
3
212
BC041114.1
-
8
652
BC051818.1
-
7
429
BC060781.1
-
8
718
4823115
BC068463.1
-
8
606
BC125104.1
-
8
547
40118340
BC125105.1
-
8
547
40118341
Back To Top
More Information
Panther Classification:
Molecular Function -
small GTPase
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
cellular homeostasis
mitochondrion transport along microtubule
regulation of small GTPase mediated signal transduction
mitochondrial outer membrane permeabilization
molecular_function
GTPase activity
calcium ion binding
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kt7sl:80/10.239.164.98:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0