Search Thermo Fisher Scientific
Gene Symbol: NUP37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024057.3 | NP_076962.2 | 4, 5 | 509 | ||
GenBank mRNA | AF514994.1 | - | 3, 4 | 363 | ||
AK026271.1 | - | 3, 4 | 344 | |||
AK296733.1 | - | 4, 5 | 467 | |||
AK310619.1 | - | 4, 5 | 469 | |||
BC000861.2 | - | 3, 4 | 427 | 3459639 | ||
DC420994.1 | - | 4, 5 | 510 |