Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FBXW12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159927.1 | NP_001153399.1 | 4 | 470 | ||
NM_001159929.1 | NP_001153401.1 | 3 | 269 | |||
NM_207102.2 | NP_996985.2 | 4 | 443 | |||
XM_017006224.1 | XP_016861713.1 | 3 | 456 | |||
GenBank mRNA | AY247969.1 | - | 4 | 443 | ||
BC101303.1 | - | 3 | 616 | 40024696 | ||
BC101304.1 | - | 4 | 414 | 40024699 | ||
BC101305.1 | - | 3 | 213 | 40024702 |