Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BAG6 SIRNA Assays ›
Gene Symbol:
BAG6
Gene Name:
BCL2 associated athanogene 6
Gene Aliases:
BAG-6, BAT3, D6S52E, G3
Chromosome Location:
Chr.6: 31639028 - 31660900 on Build GRCh38
UniGene ID:
Hs.440900
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15018
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BAG6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098534.1
NP_001092004.1
6
689
NM_001199697.1
NP_001186626.1
6
819
NM_001199698.1
NP_001186627.1
6
819
NM_004639.3
NP_004630.3
6
843
NM_080702.2
NP_542433.1
6
819
NM_080703.2
NP_542434.1
6
843
XM_011514892.2
XP_011513194.1
6
1802
XM_017011275.1
XP_016866764.1
6
1802
XM_017011276.1
XP_016866765.1
6
792
XM_017011277.1
XP_016866766.1
6
652
XM_017011278.1
XP_016866767.1
6
727
XM_017011279.1
XP_016866768.1
6
1122
XM_017011280.1
XP_016866769.1
6
694
XM_017011281.1
XP_016866770.1
6
856
XM_017011282.1
XP_016866771.1
6
648
XM_017011283.1
XP_016866772.1
6
1802
XM_017011284.1
XP_016866773.1
6
1802
XM_017011285.1
XP_016866774.1
5
592
XM_017011286.1
XP_016866775.1
5
611
XM_017011287.1
XP_016866776.1
6
1802
XM_017011288.1
XP_016866777.1
5
592
XM_017011289.1
XP_016866778.1
6
1802
XM_017011290.1
XP_016866779.1
6
1802
XM_017011291.1
XP_016866780.1
6
1802
XM_017011292.1
XP_016866781.1
6
1802
XM_017011293.1
XP_016866782.1
5
592
XM_017011294.1
XP_016866783.1
6
1802
XM_017011295.1
XP_016866784.1
6
1802
XM_017011296.1
XP_016866785.1
6
1802
XM_017011297.1
XP_016866786.1
5
592
XM_017011298.1
XP_016866787.1
6
1802
GenBank mRNA
AK302695.1
-
6
770
AK304879.1
-
6
770
BC003133.1
-
6
737
2957896
BX384328.2
-
6
822
BX647244.1
-
6
697
CX758454.1
-
6
664
M33519.1
-
6
778
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
immune system process
ubiquitin-dependent protein catabolic process
transport
synaptonemal complex assembly
spermatogenesis
brain development
embryo development
covalent chromatin modification
internal peptidyl-lysine acetylation
cell differentiation
lung development
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of cell proliferation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of apoptotic process
negative regulation of proteolysis
protein stabilization
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
tail-anchored membrane protein insertion into ER membrane
positive regulation of ERAD pathway
maintenance of unfolded protein involved in ERAD pathway
protein localization to cytosolic proteasome complex involved in ERAD pathway
protein binding
Hsp70 protein binding
polyubiquitin binding
ubiquitin protein ligase binding
ribosome binding
misfolded protein binding
proteasome binding
ubiquitin-specific protease binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-v2dhc:80/100.66.79.56:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0