Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159729.1 | NP_001153201.1 | 8 | 979 | ||
NM_008999.4 | NP_033025.2 | 8 | 1091 | |||
GenBank mRNA | AB232618.1 | - | 6 | 656 | ||
AK016493.1 | - | 8 | 991 | |||
AK019666.1 | - | 5 | 540 | |||
AK047068.1 | - | 8 | 1092 | |||
AK082017.1 | - | 8 | 1055 | |||
AK164057.1 | - | 8 | 1175 | |||
BC025578.1 | - | 8 | 986 | 5323566 | ||
Z22821.1 | - | 6 | 676 |