Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prkcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291434.1 | NP_001278363.1 | 10 | 1370 | ||
NM_011102.4 | NP_035232.1 | 11 | 1523 | |||
GenBank mRNA | AK144419.1 | - | 11 | 1525 | ||
BC111807.1 | - | 10 | 1257 | 40039740 | ||
L28035.1 | - | 11 | 1601 | |||
X67129.1 | - | 11 | 1332 |