Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKCE SIRNA Assays ›
          Gene Symbol:
          Prkce
          Gene Name:
          protein kinase C, epsilon
          Gene Aliases:
          5830406C15Rik, PKC[e], PKCepsilon, Pkce, R75156
          Chromosome Location:
          Chr.17: 86166971 - 86657919 on Build GRCm38
          UniGene ID:
          Mm.24614
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 151134

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Prkce
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011104.3 NP_035234.1 9, 10 2079
          GenBank mRNA AF028009.1 - 9, 10 1280
          AF325507.1 - 9, 10 1271
          AK082438.1 - 9, 10 2080
          AK085351.1 - 10, 11 1491

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          macrophage activation involved in immune response
          immune system process
          protein phosphorylation
          cell cycle
          cell adhesion
          chemosensory behavior
          positive regulation of epithelial cell migration
          positive regulation of fibroblast migration
          positive regulation of cell-substrate adhesion
          negative regulation of mitochondrial membrane potential
          phosphorylation
          peptidyl-serine phosphorylation
          regulation of lipid metabolic process
          positive regulation of actin filament polymerization
          lipopolysaccharide-mediated signaling pathway
          positive regulation of insulin secretion
          positive regulation of synaptic transmission, GABAergic
          positive regulation of cytokinesis
          intracellular signal transduction
          locomotory exploration behavior
          TRAM-dependent toll-like receptor 4 signaling pathway
          cellular response to platelet-derived growth factor stimulus
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to morphine
          positive regulation of MAPK cascade
          regulation of peptidyl-tyrosine phosphorylation
          positive regulation of lipid catabolic process
          release of sequestered calcium ion into cytosol
          regulation of release of sequestered calcium ion into cytosol
          negative regulation of release of sequestered calcium ion into cytosol
          cell division
          negative regulation of mitochondrial calcium ion concentration
          regulation of insulin secretion involved in cellular response to glucose stimulus
          positive regulation of mucus secretion
          cellular response to ethanol
          cellular response to prostaglandin E stimulus
          cellular response to hypoxia
          positive regulation of wound healing
          positive regulation of receptor activity
          positive regulation of cellular glucuronidation
          nucleotide binding
          actin monomer binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          calcium-independent protein kinase C activity
          receptor binding
          protein binding
          ATP binding
          enzyme activator activity
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          SH3 domain binding
          enzyme binding
          protein kinase binding
          receptor activator activity
          ethanol binding
          metal ion binding
          14-3-3 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-svdw7:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0