Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PSAP SIRNA Assays ›
          Gene Symbol:
          Psap
          Gene Name:
          prosaposin
          Gene Aliases:
          AI037048, SGP-1
          Chromosome Location:
          Chr.10: 60277628 - 60302600 on Build GRCm38
          UniGene ID:
          Mm.277498
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 151282

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Psap
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146120.1 NP_001139592.1 5, 6 691
          NM_001146121.1 NP_001139593.1 5, 6 691
          NM_001146122.1 NP_001139594.1 5, 6 691
          NM_001146123.1 NP_001139595.1 5, 6 691
          NM_001146124.1 NP_001139596.1 5, 6 607
          NM_011179.3 NP_035309.3 5, 6 691
          GenBank mRNA AK088369.1 - 5, 6 605
          AK089998.1 - 5, 6 609
          AK093881.1 - 5, 6 605
          AK135637.1 - 5, 6 605
          AK145607.1 - 5, 6 612
          AK145802.1 - 5, 6 606
          AK148150.1 - 5, 6 613
          AK148152.1 - 5, 6 632
          AK148545.1 - 5, 6 607
          AK150331.1 - 5, 6 607
          AK150645.1 - 5, 6 605
          AK151034.1 - 5, 6 605
          AK151173.1 - 5, 6 606
          AK151230.1 - 5, 6 605
          AK151234.1 - 5, 6 607
          AK151251.1 - 5, 6 605
          AK151525.1 - 5, 6 607
          AK151529.1 - 5, 6 607
          AK151686.1 - 5, 6 641
          AK151852.1 - 5, 6 607
          AK151875.1 - 5, 6 604
          AK151891.1 - 5, 6 605
          AK151907.1 - 5, 6 607
          AK151943.1 - 5, 6 607
          AK152117.1 - 5, 6 607
          AK152126.1 - 5, 6 605
          AK152134.1 - 5, 6 607
          AK152203.1 - 5, 6 639
          AK152280.1 - 5, 6 605
          AK152319.1 - 5, 6 609
          AK152355.1 - 5, 6 607
          AK152408.1 - 5, 6 610
          AK152505.1 - 5, 6 607
          AK152557.1 - 5, 6 607
          AK152605.1 - 5, 6 607
          AK152819.1 - 5, 6 619
          AK153404.1 - 5, 6 605
          AK159108.1 - 5, 6 635
          AK159164.1 - 5, 6 605
          AK159167.1 - 5, 6 606
          AK159198.1 - 5, 6 605
          AK159221.1 - 5, 6 609
          AK159244.1 - 5, 6 605
          AK159266.1 - 5, 6 605
          AK159333.1 - 5, 6 605
          AK159341.1 - 5, 6 605
          AK159414.1 - 5, 6 608
          AK159427.1 - 5, 6 607
          AK159451.1 - 5, 6 605
          AK159457.1 - 5, 6 619
          AK159463.1 - 5, 6 607
          AK159487.1 - 5, 6 605
          AK159492.1 - 5, 6 605
          AK159510.1 - 5, 6 605
          AK159528.1 - 5, 6 605
          AK159530.1 - 5, 6 605
          AK159589.1 - 5, 6 607
          AK159604.1 - 5, 6 605
          AK159622.1 - 4, 5 497
          AK159634.1 - 5, 6 605
          AK159638.1 - 5, 6 606
          AK159657.1 - 5, 6 605
          AK159664.1 - 5, 6 605
          AK159685.1 - 5, 6 605
          AK159692.1 - 5, 6 605
          AK159709.1 - 5, 6 605
          AK159720.1 - 5, 6 605
          AK159722.1 - 5, 6 605
          AK159728.1 - 5, 6 607
          AK159840.1 - 5, 6 605
          AK159861.1 - 5, 6 605
          AK159862.1 - 5, 6 607
          AK159877.1 - 5, 6 605
          AK159910.1 - 5, 6 605
          AK159915.1 - 5, 6 605
          AK159936.1 - 5, 6 607
          AK159982.1 - 5, 6 605
          AK159994.1 - 5, 6 605
          AK160015.1 - 5, 6 605
          AK160019.1 - 5, 6 607
          AK160021.1 - 5, 6 605
          AK160024.1 - 5, 6 605
          AK160046.1 - 5, 6 605
          AK160049.1 - 5, 6 607
          AK160073.1 - 5, 6 605
          AK160108.1 - 5, 6 605
          AK160877.1 - 5, 6 608
          AK161313.1 - 5, 6 622
          AK166700.1 - 5, 6 605
          AK166707.1 - 5, 6 604
          AK166744.1 - 5, 6 603
          AK166811.1 - 5, 6 605
          AK166849.1 - 5, 6 605
          AK166879.1 - 5, 6 607
          AK166909.1 - 5, 6 605
          AK166920.1 - 5, 6 605
          AK166946.1 - 5, 6 610
          AK166959.1 - 5, 6 605
          AK166984.1 - 5, 6 605
          AK167014.1 - 5, 6 605
          AK167025.1 - 5, 6 605
          AK167073.1 - 5, 6 605
          AK167099.1 - 5, 6 608
          AK167108.1 - 5, 6 605
          AK167111.1 - 5, 6 605
          AK167147.1 - 5, 6 605
          AK167198.1 - 5, 6 612
          AK167213.1 - 5, 6 605
          AK167237.1 - 5, 6 605
          AK167243.1 - 5, 6 606
          AK167252.1 - 5, 6 607
          AK167418.1 - 5, 6 607
          AK167719.1 - 5, 6 607
          AK167902.1 - 5, 6 675
          AK170416.1 - 5, 6 604
          BC036266.1 - 4, 5 471
          BC141089.1 - 5, 6 604 9055701
          BC145359.1 - 5, 6 604 9053917
          S36200.1 - 5, 6 581
          S71616.1 - 5, 6 621
          U27340.1 - 5, 6 605

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          sphingolipid metabolic process
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          regulation of lipid metabolic process
          regulation of MAPK cascade
          positive regulation of MAPK cascade
          developmental growth
          prostate gland growth
          epithelial cell differentiation involved in prostate gland development
          cellular response to organic substance
          negative regulation of hydrogen peroxide-induced cell death
          G-protein coupled receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0