Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PSAP SIRNA Assays ›
Gene Symbol:
Psap
Gene Name:
prosaposin
Gene Aliases:
AI037048, SGP-1
Chromosome Location:
Chr.10: 60277628 - 60302600 on Build GRCm38
UniGene ID:
Mm.277498
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
151284
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Psap
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146120.1
NP_001139592.1
8
901
NM_001146121.1
NP_001139593.1
9
907
NM_001146122.1
NP_001139594.1
8
901
NM_001146123.1
NP_001139595.1
8
874
NM_001146124.1
NP_001139596.1
8
817
NM_011179.3
NP_035309.3
9
910
GenBank mRNA
AK088369.1
-
8
815
AK089998.1
-
8
819
AK093881.1
-
8
815
AK135637.1
-
8
815
AK145607.1
-
8
822
AK145802.1
-
8
816
AK148150.1
-
8
823
AK148152.1
-
8
842
AK148545.1
-
9
826
AK149785.1
-
8
818
AK150331.1
-
8
817
AK150645.1
-
8
815
AK151034.1
-
8
815
AK151173.1
-
8
816
AK151230.1
-
8
815
AK151234.1
-
8
790
AK151251.1
-
8
788
AK151525.1
-
8
817
AK151529.1
-
8
790
AK151686.1
-
8
851
AK151852.1
-
8
817
AK151875.1
-
8
814
AK151891.1
-
8
815
AK151907.1
-
8
817
AK151943.1
-
8
817
AK152117.1
-
8
817
AK152126.1
-
8
815
AK152134.1
-
8
817
AK152203.1
-
8
849
AK152280.1
-
8
815
AK152319.1
-
8
819
AK152355.1
-
8
817
AK152408.1
-
8
820
AK152505.1
-
8
817
AK152557.1
-
8
817
AK152605.1
-
8
817
AK152819.1
-
8
829
AK153404.1
-
8
815
AK159108.1
-
8
845
AK159164.1
-
8
815
AK159167.1
-
8
816
AK159198.1
-
8
815
AK159221.1
-
8
819
AK159244.1
-
8
815
AK159266.1
-
8
815
AK159333.1
-
8
815
AK159341.1
-
8
815
AK159384.1
-
8
815
AK159414.1
-
8
818
AK159427.1
-
8
817
AK159451.1
-
8
815
AK159457.1
-
8
829
AK159463.1
-
8
817
AK159487.1
-
8
815
AK159492.1
-
8
815
AK159510.1
-
8
815
AK159528.1
-
8
815
AK159530.1
-
8
815
AK159589.1
-
8
817
AK159604.1
-
8
815
AK159622.1
-
7
707
AK159634.1
-
8
815
AK159638.1
-
8
816
AK159657.1
-
8
815
AK159664.1
-
8
815
AK159685.1
-
8
815
AK159692.1
-
8
815
AK159709.1
-
8
815
AK159720.1
-
8
815
AK159722.1
-
8
815
AK159728.1
-
8
817
AK159840.1
-
8
815
AK159861.1
-
8
815
AK159862.1
-
8
817
AK159877.1
-
8
815
AK159910.1
-
8
815
AK159915.1
-
8
815
AK159936.1
-
8
817
AK159982.1
-
8
815
AK159994.1
-
8
815
AK160015.1
-
8
815
AK160019.1
-
8
817
AK160021.1
-
8
815
AK160024.1
-
8
815
AK160046.1
-
8
815
AK160049.1
-
8
817
AK160073.1
-
8
815
AK160108.1
-
8
815
AK160877.1
-
8
818
AK161313.1
-
8
832
AK166700.1
-
8
815
AK166707.1
-
8
814
AK166744.1
-
8
813
AK166811.1
-
8
815
AK166849.1
-
8
815
AK166879.1
-
8
817
AK166909.1
-
8
815
AK166920.1
-
8
815
AK166946.1
-
8
820
AK166959.1
-
8
815
AK166984.1
-
8
815
AK167014.1
-
8
815
AK167025.1
-
8
815
AK167073.1
-
8
815
AK167099.1
-
8
818
AK167108.1
-
8
815
AK167111.1
-
8
815
AK167147.1
-
8
815
AK167198.1
-
8
822
AK167213.1
-
8
815
AK167237.1
-
8
815
AK167243.1
-
8
816
AK167252.1
-
8
817
AK167418.1
-
8
817
AK167719.1
-
8
817
AK167902.1
-
8
885
AK170416.1
-
8
814
AK195069.1
-
2
58
AK196371.1
-
2
58
AK201421.1
-
2
58
BC036266.1
-
7
681
BC141089.1
-
9
820
9055701
BC145359.1
-
8
814
9053917
S36200.1
-
9
800
S71616.1
-
9
840
U27340.1
-
8
815
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
sphingolipid metabolic process
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
regulation of lipid metabolic process
regulation of MAPK cascade
positive regulation of MAPK cascade
developmental growth
prostate gland growth
epithelial cell differentiation involved in prostate gland development
cellular response to organic substance
negative regulation of hydrogen peroxide-induced cell death
G-protein coupled receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0