Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163548.1 | NP_001157020.1 | 5, 6 | 893 | ||
NM_011182.4 | NP_035312.3 | 5, 6 | 569 | |||
GenBank mRNA | AB013470.1 | - | 5, 6 | 423 | ||
AF084221.1 | - | 5, 6 | 463 | |||
AK016645.1 | - | 5, 6 | 895 | |||
AK036191.1 | - | 5, 6 | 483 | |||
AK142407.1 | - | 5, 6 | 558 | |||
AK159229.1 | - | 5, 6 | 541 | |||
AK168566.1 | - | 5, 6 | 483 | |||
AK220565.1 | - | 5, 6 | 549 | |||
BC007189.1 | - | 4, 5 | 310 | |||
BC035296.1 | - | 5, 6 | 510 | 3969080 |