Search
Search

Rbl2| Gene Symbol: Rbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282000.1 | NP_001268929.1 | 7 | 985 | ||
| NM_001282001.1 | NP_001268930.1 | 8 | 1114 | |||
| NM_011250.4 | NP_035380.3 | 8 | 1114 | |||
| GenBank mRNA | AK160027.1 | - | 8 | 1061 | ||
| BC020528.1 | - | 8 | 1062 | 4240592 | ||
| U36799.1 | - | 8 | 1014 | |||
| U47333.1 | - | 7 | 927 | |||
| U50850.1 | - | 8 | 1022 | |||