Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNAI2 SIRNA Assays ›
Gene Symbol:
Snai2
Gene Name:
snail family zinc finger 2
Gene Aliases:
Slug, Slugh, Snail2
Chromosome Location:
Chr.16: 14705859 - 14709385 on Build GRCm38
UniGene ID:
Mm.4272
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
151769
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Snai2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_011415.2
NP_035545.1
3
943
GenBank mRNA
AK133968.1
-
3
951
BC062164.1
-
3
943
30242610
U79550.1
-
3
871
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
epithelial to mesenchymal transition
epithelial to mesenchymal transition involved in endocardial cushion formation
cell migration involved in endocardial cushion formation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
negative regulation of cell adhesion involved in substrate-bound cell migration
Notch signaling pathway
multicellular organism development
sensory perception of sound
response to radiation
negative regulation of keratinocyte proliferation
negative regulation of vitamin D biosynthetic process
neural crest cell development
cell migration
positive regulation of cell migration
negative regulation of chondrocyte differentiation
regulation of chemokine production
positive regulation of histone acetylation
negative regulation of catenin import into nucleus
desmosome disassembly
cellular response to platelet-derived growth factor stimulus
regulation of apoptotic process
negative regulation of apoptotic process
pigmentation
negative regulation of DNA damage response, signal transduction by p53 class mediator
wound healing, spreading of cells
positive regulation of fat cell differentiation
regulation of osteoblast differentiation
white fat cell differentiation
palate development
canonical Wnt signaling pathway
epithelium development
cartilage morphogenesis
regulation of branching involved in salivary gland morphogenesis
negative regulation of vitamin D receptor signaling pathway
cellular response to epidermal growth factor stimulus
cellular response to ionizing radiation
negative regulation of canonical Wnt signaling pathway
negative regulation of cell-cell adhesion by negative regulation of transcription from RNA polymerase II promoter
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of stem cell proliferation
regulation of bicellular tight junction assembly
negative regulation of anoikis
positive regulation of endothelial cell chemotaxis
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mc8hj:80/100.66.75.242:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0