Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POU5F1 SIRNA Assays ›
          Gene Symbol:
          Pou5f1 Gm5712
          Gene Name:
          POU domain, class 5, transcription factor 1
          POU domain, class 5, transcription factor 1 pseudogene
          Gene Aliases:
          NF-A3, Oct-3, Oct-3/4, Oct-4, Oct3, Oct3/4, Oct4, Otf-3, Otf-4, Otf3, Otf3-rs7, Otf3g, Otf4
          EG435755
          Chromosome Location:
          Chr.17: 35506032 - 35510777 on Build GRCm38
          Chromosome Location:
          Chr.3: 129433673 - 129434712 on Build GRCm38
          UniGene ID:
          Mm.17031
          Mm.430561
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 151958

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pou5f1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252452.1 NP_001239381.1 3, 4 341
          NM_013633.3 NP_038661.2 3, 4 703
          GenBank mRNA AB221654.1 - 3, 4 635
          AB375269.1 - 3, 4 280
          AB375270.1 - 3, 4 341
          AB375271.1 - 2, 3 533
          AB375272.1 - 3, 4 307
          AB375273.1 - 3, 4 322
          AB375274.1 - 1, 2 153
          AB375275.1 - 14
          AB375276.1 - 3, 4 837
          AB375277.1 - 2, 3 299
          AK145321.1 - 3, 4 674
          BC068268.1 - 3, 4 679 30019896
          M34381.1 - 3, 4 684
          X52437.1 - 3, 4 582
          Gene Symbol: Gm5712
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA DQ851563.1 - 1 524
          DQ851564.1 - 1 608
          EF187444.1 - 1 525

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: S81255.
          This siRNA also targets the following transcripts in Rat: NM_001009178, BC158566.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          mesodermal cell fate commitment
          endodermal cell fate commitment
          ectodermal cell fate commitment
          endodermal cell fate specification
          trophectodermal cell differentiation
          blastocyst growth
          BMP signaling pathway involved in heart induction
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          multicellular organism development
          response to organic substance
          regulation of gene expression
          stem cell population maintenance
          germ-line stem cell population maintenance
          response to retinoic acid
          somatic stem cell population maintenance
          positive regulation of catenin import into nucleus
          mRNA transcription from RNA polymerase II promoter
          cell fate commitment
          negative regulation of cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of calcium ion-dependent exocytosis
          stem cell differentiation
          positive regulation of protein kinase B signaling
          negative regulation of protein kinase B signaling
          positive regulation of SMAD protein import into nucleus
          cell fate commitment involved in formation of primary germ layer
          cardiac cell fate determination
          negative regulation of gene silencing by miRNA
          regulation of heart induction by regulation of canonical Wnt signaling pathway
          regulation of methylation-dependent chromatin silencing
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription coactivator activity
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          transcription factor binding
          chromatin DNA binding
          ubiquitin protein ligase binding
          miRNA binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          poly(A) RNA binding
          protein heterodimerization activity
          POU domain binding
          HMG box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4xjcb:80/100.66.78.128:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0