Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172152.1 | NP_001165623.1 | 2 | 170 | ||
NM_028220.3 | NP_082496.3 | 3 | 244 | |||
GenBank mRNA | AK013889.1 | - | 2 | 150 | ||
AK020995.1 | - | 3 | 267 | |||
AK044241.1 | - | 2 | 171 | |||
BC028268.1 | - | 3 | 255 | 5366098 |