Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARHGEF2 SIRNA Assays ›
          Gene Symbol:
          ARHGEF2
          Gene Name:
          Rho/Rac guanine nucleotide exchange factor 2
          Gene Aliases:
          GEF, GEF-H1, GEFH1, LFP40, P40
          Chromosome Location:
          Chr.1: 155946839 - 155991263 on Build GRCh38
          UniGene ID:
          Hs.743352
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 15256

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARHGEF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001162383.1 NP_001155855.1 3, 4 390
          NM_001162384.1 NP_001155856.1 3, 4 390
          NM_004723.3 NP_004714.2 3, 4 318
          XM_005245587.2 XP_005245644.1 3, 4 443
          XM_005245588.2 XP_005245645.1 3, 4 443
          XM_005245589.4 XP_005245646.1 3, 4 596
          XM_005245590.3 XP_005245647.1 3, 4 318
          XM_005245591.3 XP_005245648.1 4, 5 434
          XM_005245592.2 XP_005245649.1 3, 4 587
          XM_005245593.4 XP_005245650.1 3, 4 318
          XM_005245594.2 XP_005245651.1 3, 4 1304
          XM_006711622.3 XP_006711685.1 3, 4 1581
          XM_011510136.2 XP_011508438.2 7, 8 1002
          XM_011510137.2 XP_011508439.2 7, 8 1002
          XM_011510139.2 XP_011508441.1 7, 8 1002
          XM_011510141.2 XP_011508443.1 3, 4 1120
          XM_017002794.1 XP_016858283.1 7, 8 1002
          XM_017002795.1 XP_016858284.1 3, 4 337
          XM_017002796.1 XP_016858285.1 3, 4 1581
          XM_017002797.1 XP_016858286.1 3, 4 321
          GenBank mRNA AB014551.3 - 7, 8 661
          AF486838.1 - 3, 4 272
          AK295924.1 - 3, 4 587
          AK304548.1 - 6, 7 727
          AL512715.1 - 3, 4 303
          BC020567.1 - 3, 4 372 4157775
          BT007407.1 - 2, 3 191
          CN388788.1 - 3, 4 392

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001012079, XM_006232643, XM_006232648, XM_006232652, XM_008761152, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001198912, NM_001198911, NM_008487, NM_001198913, AF177032, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          cell morphogenesis
          intracellular protein transport
          actin filament organization
          negative regulation of microtubule depolymerization
          mitotic nuclear division
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          regulation of Rho protein signal transduction
          intracellular signal transduction
          regulation of cell proliferation
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          innate immune response
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of neurogenesis
          regulation of small GTPase mediated signal transduction
          positive regulation of NF-kappaB transcription factor activity
          cell division
          negative regulation of necroptotic process
          cellular response to muramyl dipeptide
          cellular response to tumor necrosis factor
          cellular hyperosmotic response
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          protein binding
          microtubule binding
          transcription factor binding
          zinc ion binding
          Rho GTPase binding
          Rac guanyl-nucleotide exchange factor activity
          Rac GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0