Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SULF1 SIRNA Assays ›
          Gene Symbol:
          Sulf1
          Gene Name:
          sulfatase 1
          Gene Aliases:
          AI467640, AW121680, mKIAA1077
          Chromosome Location:
          Chr.1: 12691695 - 12861192 on Build GRCm38
          UniGene ID:
          Mm.45563
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 153124

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sulf1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198565.1 NP_001185494.1 5, 6 902
          NM_001198566.1 NP_001185495.1 4, 5 807
          NM_172294.1 NP_758498.1 5, 6 886
          GenBank mRNA AK028285.1 - 4, 5 706
          AK045002.1 - 5, 6 801
          AK078756.1 - 5, 6 891
          AK129278.1 - 4, 5 708
          AY101178.1 - 5, 6 911
          BC034547.1 - 5, 6 886 4500954
          BC049276.1 - 5, 6 886 4500954

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_015170, NM_001128204, NM_001128205, NM_001128206, AF545571, and others.
          This siRNA also targets the following transcripts in Rat: NM_134378, XM_008763487, XM_008763488, XM_008763486, AF230072.

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          negative regulation of endothelial cell proliferation
          chondrocyte development
          glomerular filtration
          sulfur compound metabolic process
          apoptotic process
          metabolic process
          positive regulation of vascular endothelial growth factor production
          esophagus smooth muscle contraction
          negative regulation of angiogenesis
          positive regulation of Wnt signaling pathway
          heparan sulfate proteoglycan metabolic process
          negative regulation of cell migration
          positive regulation of BMP signaling pathway
          glomerular basement membrane development
          glial cell-derived neurotrophic factor receptor signaling pathway
          regulation of fibroblast growth factor receptor signaling pathway
          negative regulation of fibroblast growth factor receptor signaling pathway
          vascular endothelial growth factor receptor signaling pathway
          embryonic skeletal system development
          cartilage development
          bone development
          innervation
          negative regulation of prostatic bud formation
          catalytic activity
          arylsulfatase activity
          calcium ion binding
          N-acetylglucosamine-6-sulfatase activity
          sulfuric ester hydrolase activity
          hydrolase activity
          alkyl sulfatase activity
          endosulfan hemisulfate sulfatase activity
          endosulfan sulfate hydrolase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0