Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175552.4 | NP_780761.1 | 3 | 435 | ||
GenBank mRNA | AK043557.1 | - | 3 | 368 | ||
AK050738.1 | - | 3 | 403 | |||
AK050797.1 | - | 3 | 406 | |||
BC063100.1 | - | 3 | 421 | 6332749 |