Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183029.2 | NP_898850.2 | 6 | 967 | ||
GenBank mRNA | AK044984.1 | - | 5 | 483 | ||
AK049196.1 | - | 6 | 664 | |||
AK077404.1 | - | 6 | 647 | |||
AK159654.1 | - | 6 | 700 | |||
AK170531.1 | - | 5 | 471 | |||
AY531659.1 | - | 6 | 631 | |||
BC023758.1 | - | 6 | 665 | |||
BC054552.1 | - | 6 | 687 |