Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Orai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198424.3 | NP_940816.1 | 2 | 1543 | ||
GenBank mRNA | AK037138.1 | - | 3 | 1687 | ||
AK136955.1 | - | 2 | 1544 | |||
AK208652.1 | - | 1 | 251 | |||
BC061259.1 | - | 2 | 1513 | 30251946 |