Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JAK2 SIRNA Assays ›
          Gene Symbol:
          Jak2
          Gene Name:
          Janus kinase 2
          Gene Aliases:
          Fd17
          Chromosome Location:
          Chr.19: 29251776 - 29313080 on Build GRCm38
          UniGene ID:
          Mm.275839
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 154961

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Jak2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001048177.2 NP_001041642.1 11 1932
          NM_008413.3 NP_032439.2 12 2015
          GenBank mRNA AK087255.1 - 11 1899
          BC054807.1 - 11 1695 6838318
          BC059834.1 - 11 1695 6838318
          L16956.1 - 11 1684

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031514, AJ000557, U13396.

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          immune system process
          protein phosphorylation
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          response to oxidative stress
          signal transduction
          enzyme linked receptor protein signaling pathway
          transmembrane receptor protein tyrosine kinase signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          JAK-STAT cascade
          tyrosine phosphorylation of STAT protein
          STAT protein import into nucleus
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to oxidative stress
          hormone-mediated signaling pathway
          negative regulation of cardiac muscle cell apoptotic process
          positive regulation of cell-substrate adhesion
          positive regulation of phosphatidylinositol 3-kinase signaling
          phosphorylation
          cell migration
          chromatin modification
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          negative regulation of cell-cell adhesion
          myeloid cell differentiation
          cell differentiation
          regulation of cell adhesion
          erythrocyte differentiation
          positive regulation of cell migration
          axon regeneration
          mineralocorticoid receptor signaling pathway
          positive regulation of insulin secretion
          response to lipopolysaccharide
          positive regulation of phosphoprotein phosphatase activity
          positive regulation of interleukin-1 beta production
          positive regulation of tumor necrosis factor production
          positive regulation of protein import into nucleus, translocation
          response to hydroperoxide
          tumor necrosis factor-mediated signaling pathway
          host programmed cell death induced by symbiont
          response to tumor necrosis factor
          histone H3-Y41 phosphorylation
          intracellular signal transduction
          interleukin-12-mediated signaling pathway
          peptidyl-tyrosine autophosphorylation
          tyrosine phosphorylation of Stat3 protein
          tyrosine phosphorylation of Stat5 protein
          tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of tyrosine phosphorylation of Stat5 protein
          activation of JAK2 kinase activity
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of DNA binding
          negative regulation of DNA binding
          negative regulation of neuron apoptotic process
          innate immune response
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cell differentiation
          negative regulation of heart contraction
          positive regulation of transcription from RNA polymerase II promoter
          response to antibiotic
          protein autophosphorylation
          platelet-derived growth factor receptor signaling pathway
          regulation of inflammatory response
          positive regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of cell activation
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of nitric-oxide synthase biosynthetic process
          interferon-gamma-mediated signaling pathway
          growth hormone receptor signaling pathway
          JAK-STAT cascade involved in growth hormone signaling pathway
          positive regulation of growth hormone receptor signaling pathway
          mammary gland epithelium development
          response to interleukin-12
          cellular response to lipopolysaccharide
          cellular response to dexamethasone stimulus
          response to granulocyte macrophage colony-stimulating factor
          extrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          positive regulation of apoptotic signaling pathway
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor binding
          growth hormone receptor binding
          interleukin-12 receptor binding
          protein binding
          ATP binding
          protein C-terminus binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein kinase binding
          heme binding
          type 1 angiotensin receptor binding
          acetylcholine receptor binding
          histone kinase activity (H3-Y41 specific)
          SH2 domain binding
          histone binding
          phosphatidylinositol 3-kinase binding
          insulin receptor substrate binding
          peptide hormone receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0