Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008610.2 | NP_032636.1 | 9, 10 | 1778 | ||
GenBank mRNA | AK148184.1 | - | 9, 10 | 1777 | ||
AY622966.1 | - | 2, 3 | 207 | |||
BC070430.1 | - | 9, 10 | 1778 | 6813184 | ||
M84324.1 | - | 9, 10 | 1524 |