Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Msi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008629.1 | NP_032655.1 | 8 | 547 | ||
GenBank mRNA | AK084019.1 | - | 8 | 530 | ||
D49654.1 | - | 8 | 547 |