Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Itih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010582.3 | NP_034712.2 | 10 | 1219 | ||
GenBank mRNA | AK133782.1 | - | 10 | 1118 | ||
AK149533.1 | - | 11 | 1264 | |||
BC034341.1 | - | 10 | 1193 | 4211576 | ||
X70392.1 | - | 10 | 1205 |