Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JAK3 SIRNA Assays ›
          Gene Symbol:
          Jak3
          Gene Name:
          Janus kinase 3
          Gene Aliases:
          fae, wil
          Chromosome Location:
          Chr.8: 71676383 - 71690577 on Build GRCm38
          UniGene ID:
          Mm.249645
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 155938

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Jak3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190830.1 NP_001177759.1 12 1793
          NM_010589.6 NP_034719.2 12 1793
          GenBank mRNA AK038268.1 - 11 1604
          AK088365.1 - 12 1642
          AK142178.1 - 12 1631
          AK156646.1 - 12 1615
          AK164520.1 - 12 1633
          BC105577.1 - 11 1596 3968386
          BC131646.1 - 11 1604 40110146
          BC131647.1 - 11 1604 40110148
          L32955.1 - 11 1592
          L33768.1 - 9 2234
          L40172.1 - 12 1654

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          negative regulation of dendritic cell cytokine production
          protein phosphorylation
          enzyme linked receptor protein signaling pathway
          transmembrane receptor protein tyrosine kinase signaling pathway
          positive regulation of cytosolic calcium ion concentration
          tyrosine phosphorylation of STAT protein
          phosphorylation
          cell migration
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          B cell differentiation
          erythrocyte differentiation
          negative regulation of interleukin-10 production
          negative regulation of interleukin-12 production
          intracellular signal transduction
          interleukin-4-mediated signaling pathway
          peptidyl-tyrosine autophosphorylation
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          tyrosine phosphorylation of Stat5 protein
          T cell homeostasis
          innate immune response
          negative regulation of FasL biosynthetic process
          negative regulation of T-helper 1 cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          lymph node development
          positive regulation of immune response
          negative regulation of T cell activation
          positive regulation of calcium ion transport
          regulation of T cell apoptotic process
          negative regulation of thymocyte apoptotic process
          response to interleukin-2
          response to interleukin-4
          response to interleukin-15
          response to interleukin-9
          positive regulation of dendritic cell apoptotic process
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein phosphatase binding
          hormone receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ndjsg:80/100.66.79.104:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0