Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NCK1 SIRNA Assays ›
Gene Symbol:
Nck1
Gene Name:
non-catalytic region of tyrosine kinase adaptor protein 1
Gene Aliases:
6330586M15Rik, D230010O13Rik, Nck, Nck-1
Chromosome Location:
Chr.9: 100494981 - 100546053 on Build GRCm38
UniGene ID:
Mm.181485
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
156448
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nck1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_010878.2
NP_035008.2
2, 3
301
GenBank mRNA
AF084183.1
-
1, 2
224
BC002015.1
-
2, 3
301
3593441
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translation
negative regulation of protein kinase activity
substrate-dependent cell migration, cell extension
actin filament organization
cell migration
lamellipodium assembly
positive regulation of actin filament polymerization
negative regulation of peptidyl-serine phosphorylation
positive regulation of translation in response to endoplasmic reticulum stress
positive regulation of T cell proliferation
T cell activation
positive regulation of transcription from RNA polymerase II promoter
response to other organism
negative regulation of cell death
peptidyl-serine dephosphorylation
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of cap-dependent translational initiation
positive regulation of cap-independent translational initiation
negative regulation of PERK-mediated unfolded protein response
negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
protein kinase inhibitor activity
receptor binding
protein binding
protein kinase regulator activity
protein domain specific binding
receptor tyrosine kinase binding
eukaryotic initiation factor eIF2 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0