Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ly75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013825.3 | NP_038853.2 | 11 | 1619 | ||
GenBank mRNA | AF395445.1 | - | 11 | 1619 | ||
AK144922.1 | - | 11 | 1672 | |||
AK157921.1 | - | 11 | 1683 | |||
BC150734.1 | - | 1640 | 9087645 | |||
U19271.1 | - | 1653 |