Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATXN3 SIRNA Assays ›
          Gene Symbol:
          ATXN3 ATXN8
          Gene Name:
          ataxin 3
          ataxin 8
          Gene Aliases:
          AT3, ATX3, JOS, MJD, MJD1, SCA3
          ATXN3
          Chromosome Location:
          Chr.14: 92058552 - 92106621 on Build GRCh38
          Chromosome Location:
          -
          UniGene ID:
          Hs.532632
          Hs.532632
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 15691

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATXN3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127696.1 NP_001121168.1 7, 8 807
          NM_001127697.2 NP_001121169.2 6, 7 699
          NM_001164779.1 NP_001158251.1 4, 5 489
          NM_001164780.1 NP_001158252.1 5, 6 556
          NM_001164781.1 NP_001158253.1 6, 7 642
          NM_004993.5 NP_004984.2 8, 9 852
          NM_030660.4 NP_109376.1 7, 8 687
          NR_028453.1 7, 8 766
          NR_028454.1 6, 7 601
          NR_028455.1 7, 8 766
          NR_028456.1 6, 7 601
          NR_028457.1 8, 9 857
          NR_028458.1 7, 8 743
          NR_028459.1 8, 9 852
          NR_028461.1 7, 8 764
          NR_028462.1 7, 8 743
          NR_028463.1 3, 4 401
          NR_028464.1 6, 7 699
          NR_028465.1 6, 7 721
          NR_028466.1 3, 4 401
          NR_028467.1 7, 8 767
          NR_028468.1 6, 7 599
          NR_028469.1 6, 7 601
          NR_031765.1 2, 3 268
          GenBank mRNA AB050194.1 - 7, 8 676
          AB209309.1 - 1, 2 128
          AK225884.1 - 8, 9 791
          AK307720.1 - 5, 6 517
          AK314919.1 - 8, 9 804
          BC022245.1 - 7, 8 735
          BC033711.1 - 8, 9 795 4393766
          BC095402.1 - 8, 9 812 30347215
          BU190081.1 - 7, 8 786
          BU663113.1 - 5, 6 606
          GQ176433.1 - 8, 9 805
          GQ176434.1 - 8, 9 804
          GQ176435.1 - 8, 9 805
          GQ176436.1 - 6, 7 652
          GQ176437.1 - 6, 7 652
          GQ176438.1 - 7, 8 719
          GQ176440.1 - 7, 8 640
          GQ176441.1 - 7, 8 719
          GQ176442.1 - 6, 7 554
          GQ176443.1 - 7, 8 719
          GQ176445.1 - 6, 7 554
          GQ176448.1 - 8, 9 805
          GQ176451.1 - 6, 7 554
          GQ176452.1 - 7, 8 805
          GQ176453.1 - 7, 8 695
          GQ176454.1 - 8, 9 805
          GQ176455.1 - 7, 8 640
          GQ176456.1 - 6, 7 554
          GQ176457.1 - 6, 7 652
          GQ176458.1 - 7, 8 719
          GQ176459.1 - 4, 5 442
          GQ176462.1 - 8, 9 805
          GQ176463.1 - 7, 8 719
          GQ176464.1 - 7, 8 719
          GQ176465.1 - 3, 4 354
          GQ176466.1 - 6, 7 554
          GQ176467.1 - 7, 8 640
          GQ176469.1 - 6, 7 652
          GQ176470.1 - 6, 7 674
          GQ176474.1 - 6, 7 652
          GQ176475.1 - 6, 7 554
          GQ176476.1 - 8, 9 805
          GQ176477.1 - 7, 8 719
          GQ176479.1 - 7, 8 719
          GQ176480.1 - 6, 7 552
          GQ176481.1 - 7, 8 719
          GQ176483.1 - 7, 8 719
          GQ176484.1 - 7, 8 719
          GQ176485.1 - 6, 7 651
          GQ176486.1 - 7, 8 760
          GQ176487.1 - 6, 7 652
          GQ176488.1 - 8, 9 805
          GQ176489.1 - 7, 8 760
          GQ176490.1 - 6, 7 686
          GQ176492.1 - 7, 8 719
          GQ176493.1 - 6, 7 651
          GQ176495.1 - 6, 7 652
          GQ176496.1 - 6, 7 554
          GQ176497.1 - 7, 8 719
          GQ176498.1 - 7, 8 640
          GQ176499.1 - 8, 9 805
          GQ176500.1 - 8, 9 806
          GQ176501.1 - 7, 8 696
          GQ176503.1 - 6, 7 553
          GQ176505.1 - 6, 7 554
          GQ176506.1 - 6, 7 554
          GQ176508.1 - 8, 9 805
          GQ176509.1 - 6, 7 554
          GQ176510.1 - 8, 9 805
          GQ176511.1 - 6, 7 652
          GQ176512.1 - 8, 9 802
          GQ176513.1 - 7, 8 719
          GQ176514.1 - 5, 6 509
          GQ176515.1 - 6, 7 552
          GQ176516.1 - 8, 9 805
          GQ176518.1 - 3, 4 354
          GQ176519.1 - 7, 8 718
          GQ176520.1 - 5, 6 509
          GQ176521.1 - 6, 7 554
          GQ176522.1 - 6, 7 554
          GQ176523.1 - 8, 9 805
          GQ176524.1 - 7, 8 719
          GQ176526.1 - 6, 7 595
          GQ176527.1 - 7, 8 719
          GQ176528.1 - 7, 8 719
          GQ176529.1 - 7, 8 716
          GQ176530.1 - 7, 8 719
          GQ176531.1 - 8, 9 805
          GQ176532.1 - 7, 8 805
          GQ176533.1 - 8, 9 805
          GQ176536.1 - 7, 8 640
          GQ176537.1 - 7, 8 719
          GQ176540.1 - 7, 8 693
          GQ176542.1 - 4, 5 441
          GQ176543.1 - 6, 7 554
          GQ176544.1 - 6, 7 554
          GQ176545.1 - 4, 5 442
          GQ176546.1 - 7, 8 719
          GQ176547.1 - 6, 7 554
          GQ176548.1 - 8, 9 805
          GQ176549.1 - 6, 7 554
          GQ176550.1 - 8, 9 805
          GQ176551.1 - 8, 9 803
          GQ176552.1 - 8, 9 804
          GQ176553.1 - 6, 7 552
          GQ176554.1 - 6, 7 554
          GQ176556.1 - 6, 7 554
          GQ176558.1 - 6, 7 552
          GQ176560.1 - 8, 9 805
          GQ176563.1 - 8, 9 805
          GQ176565.1 - 8, 9 805
          GQ176566.1 - 7, 8 640
          GQ176567.1 - 8, 9 805
          GQ176568.1 - 8, 9 805
          GQ176569.1 - 7, 8 716
          GQ176570.1 - 8, 9 805
          GQ176571.1 - 6, 7 554
          GQ176572.1 - 7, 8 719
          GQ176576.1 - 7, 8 717
          GQ176577.1 - 8, 9 805
          GQ176578.1 - 8, 9 805
          GQ176579.1 - 7, 8 696
          GQ176580.1 - 8, 9 805
          GQ176583.1 - 2, 3 221
          GQ176586.1 - 7, 8 719
          GQ176587.1 - 6, 7 554
          GQ176588.1 - 7, 8 719
          GQ176590.1 - 2, 3 221
          GQ176591.1 - 6, 7 554
          GQ176592.1 - 7, 8 639
          GQ176593.1 - 9, 10 891
          GQ176594.1 - 8, 9 805
          GQ176595.1 - 6, 7 552
          GQ176596.1 - 7, 8 719
          GQ176598.1 - 6, 7 554
          GQ176599.1 - 8, 9 805
          GQ176600.1 - 7, 8 719
          GQ176601.1 - 6, 7 554
          GQ176602.1 - 8, 9 823
          GQ176603.1 - 6, 7 554
          GQ176604.1 - 6, 7 554
          GQ176605.1 - 7, 8 719
          GQ176607.1 - 7, 8 719
          GQ176608.1 - 6, 7 652
          GQ176609.1 - 805
          GQ176610.1 - 7, 8 719
          GQ176611.1 - 7, 8 719
          GQ176612.1 - 8, 9 805
          GQ176613.1 - 3, 4 331
          GQ176614.1 - 5, 6 509
          GQ176615.1 - 805
          GQ176616.1 - 8, 9 805
          GQ176617.1 - 7, 8 719
          GQ176618.1 - 631
          GQ176619.1 - 8, 9 805
          GQ176620.1 - 7, 8 719
          GQ176621.1 - 8, 9 805
          GQ176622.1 - 8, 9 805
          GQ176624.1 - 7, 8 738
          GQ176625.1 - 442
          GQ176626.1 - 6, 7 554
          GQ176627.1 - 4, 5 442
          GQ176628.1 - 805
          GQ176629.1 - 719
          GQ176630.1 - 7, 8 719
          GQ176631.1 - 6, 7 652
          GQ176632.1 - 8, 9 805
          GQ176633.1 - 4, 5 442
          GQ176635.1 - 554
          GQ176636.1 - 5, 6 626
          GQ176637.1 - 6, 7 554
          GQ176638.1 - 6, 7 554
          GQ176639.1 - 6, 7 651
          GQ176640.1 - 8, 9 805
          GQ176641.1 - 525
          GQ176642.1 - 8, 9 805
          GQ176643.1 - 554
          GQ176644.1 - 8, 9 800
          GQ176645.1 - 8, 9 805
          GQ176646.1 - 8, 9 805
          GQ176647.1 - 554
          GQ176649.1 - 7, 8 719
          GQ176650.1 - 3, 4 354
          GQ176653.1 - 5, 6 509
          GQ176654.1 - 7, 8 719
          GQ176655.1 - 6, 7 649
          GQ176656.1 - 6, 7 647
          GQ176657.1 - 6, 7 554
          GQ176658.1 - 8, 9 805
          GQ176659.1 - 719
          GQ176660.1 - 7, 8 719
          GQ176661.1 - 5, 6 509
          GQ176662.1 - 7, 8 719
          GQ176663.1 - 6, 7 652
          GQ176664.1 - 7, 8 719
          GQ176665.1 - 7, 8 640
          GQ176666.1 - 5, 6 509
          GQ176668.1 - 8, 9 802
          GQ176669.1 - 719
          GQ176670.1 - 5, 6 607
          GQ176671.1 - 805
          GQ176673.1 - 8, 9 805
          GQ176674.1 - 8, 9 805
          GQ176675.1 - 7, 8 640
          GQ176676.1 - 6, 7 554
          GQ176677.1 - 8, 9 805
          GQ176678.1 - 7, 8 719
          GQ176679.1 - 7, 8 640
          GQ176680.1 - 8, 9 805
          GQ176681.1 - 5, 6 564
          GQ176682.1 - 7, 8 719
          GQ176685.1 - 8, 9 804
          GQ176687.1 - 8, 9 805
          GQ176688.1 - 6, 7 651
          GQ176689.1 - 5, 6 509
          GQ176690.1 - 7, 8 719
          GQ176691.1 - 6, 7 554
          GQ176692.1 - 8, 9 805
          GQ176693.1 - 805
          GQ176694.1 - 6, 7 473
          GQ176695.1 - 6, 7 652
          GQ176696.1 - 7, 8 719
          GQ176697.1 - 8, 9 805
          GQ176698.1 - 805
          GQ176699.1 - 805
          GQ176700.1 - 7, 8 640
          GQ176701.1 - 8, 9 805
          GQ176702.1 - 5, 6 509
          GQ176703.1 - 8, 9 805
          GQ176706.1 - 7, 8 719
          GQ176707.1 - 551
          GQ176708.1 - 9, 10 891
          GQ176709.1 - 716
          GQ176710.1 - 4, 5 442
          GQ176711.1 - 5, 6 509
          GQ176712.1 - 7, 8 760
          GQ176713.1 - 554
          GQ176714.1 - 8, 9 805
          GQ176715.1 - 8, 9 727
          GQ176716.1 - 6, 7 554
          GQ176717.1 - 719
          GQ176719.1 - 805
          GQ176720.1 - 6, 7 554
          GQ176721.1 - 8, 9 805
          GQ176722.1 - 7, 8 719
          GQ176723.1 - 5, 6 509
          GQ176724.1 - 6, 7 652
          GQ176725.1 - 8, 9 805
          GQ176726.1 - 6, 7 554
          GQ176728.1 - 8, 9 805
          GQ176731.1 - 6, 7 554
          GQ176732.1 - 554
          GQ176734.1 - 652
          GQ176735.1 - 8, 9 805
          GQ176736.1 - 554
          GQ176737.1 - 8, 9 805
          GQ176738.1 - 7, 8 719
          GQ176740.1 - 6, 7 554
          GQ176741.1 - 805
          GQ176743.1 - 6, 7 554
          GQ176744.1 - 7, 8 760
          GQ176746.1 - 8, 9 805
          GQ176747.1 - 805
          GQ176748.1 - 6, 7 554
          GQ176749.1 - 8, 9 805
          GQ176750.1 - 7, 8 640
          GQ176751.1 - 554
          GQ176753.1 - 5, 6 509
          GQ176754.1 - 7, 8 640
          GQ176755.1 - 7, 8 719
          GQ176756.1 - 8, 9 805
          GQ176757.1 - 6, 7 652
          GQ176758.1 - 6, 7 554
          GQ176759.1 - 8, 9 805
          GQ176760.1 - 7, 8 719
          GQ176761.1 - 5, 6 487
          GQ176762.1 - 805
          GQ176763.1 - 8, 9 805
          GQ176765.1 - 6, 7 554
          GQ176766.1 - 8, 9 805
          GQ176767.1 - 7, 8 719
          GQ176768.1 - 6, 7 554
          GQ176769.1 - 5, 6 607
          GQ176770.1 - 5, 6 387
          GQ176771.1 - 8, 9 805
          GQ176772.1 - 6, 7 652
          GQ176773.1 - 6, 7 652
          GQ176774.1 - 6, 7 554
          GQ176776.1 - 805
          GQ176778.1 - 5, 6 509
          GQ176779.1 - 6, 7 652
          GQ176780.1 - 6, 7 554
          GQ176781.1 - 6, 7 652
          GQ176782.1 - 6, 7 652
          GQ176783.1 - 554
          GQ176784.1 - 6, 7 652
          GQ176785.1 - 5, 6 487
          GQ176786.1 - 6, 7 652
          GQ176787.1 - 8, 9 805
          GQ176788.1 - 6, 7 554
          GQ176789.1 - 6, 7 651
          GQ176791.1 - 8, 9 805
          GQ176792.1 - 6, 7 649
          GQ176794.1 - 509
          GQ176795.1 - 6, 7 554
          GQ176797.1 - 7, 8 719
          GQ176798.1 - 719
          GQ176799.1 - 487
          GQ176800.1 - 805
          GQ176801.1 - 718
          GQ176802.1 - 652
          GQ176803.1 - 719
          GQ176804.1 - 442
          GQ176805.1 - 8, 9 781
          GQ176806.1 - 8, 9 804
          GQ176807.1 - 7, 8 719
          GQ176808.1 - 805
          GQ176810.1 - 7, 8 717
          GQ176811.1 - 8, 9 806
          GQ176812.1 - 6, 7 652
          GQ176813.1 - 7, 8 640
          GQ176814.1 - 6, 7 551
          GQ176815.1 - 6, 7 653
          GQ176816.1 - 719
          GQ176817.1 - 4, 5 441
          GQ176818.1 - 7, 8 760
          GQ176819.1 - 7, 8 719
          GQ176820.1 - 8, 9 805
          GQ176821.1 - 6, 7 553
          GQ176822.1 - 7, 8 640
          GQ176826.1 - 8, 9 782
          GQ176827.1 - 8, 9 805
          GQ176828.1 - 6, 7 554
          GQ176829.1 - 8, 9 805
          GQ176830.1 - 776
          GQ176831.1 - 805
          GQ176832.1 - 6, 7 554
          GQ176834.1 - 718
          GQ176835.1 - 7, 8 639
          GQ176836.1 - 7, 8 639
          GQ176837.1 - 8, 9 805
          GQ176838.1 - 8, 9 805
          GQ176840.1 - 6, 7 554
          GQ176841.1 - 8, 9 805
          GQ176842.1 - 719
          GQ176843.1 - 6, 7 553
          GQ176844.1 - 6, 7 567
          GQ176845.1 - 7, 8 718
          GQ176846.1 - 8, 9 805
          GQ176847.1 - 6, 7 551
          S75313.1 - 8, 9 818
          U64820.1 - 8, 9 840
          U64821.1 - 8, 9 803
          U64822.1 - 7, 8 780
          Gene Symbol: ATXN8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA GQ176777.1 - 487

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          nucleotide-excision repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          ubiquitin-dependent protein catabolic process
          misfolded or incompletely synthesized protein catabolic process
          chemical synaptic transmission
          nervous system development
          regulation of cell-substrate adhesion
          protein deubiquitination
          actin cytoskeleton organization
          cellular response to heat
          monoubiquitinated protein deubiquitination
          exploration behavior
          proteasome-mediated ubiquitin-dependent protein catabolic process
          intermediate filament cytoskeleton organization
          protein K63-linked deubiquitination
          histone H3 deacetylation
          protein K48-linked deubiquitination
          cellular response to misfolded protein
          positive regulation of ERAD pathway
          protein localization to cytosolic proteasome complex involved in ERAD pathway
          RNA polymerase II regulatory region DNA binding
          histone deacetylase activity
          thiol-dependent ubiquitin-specific protease activity
          protein binding
          omega peptidase activity
          ubiquitin protein ligase binding
          identical protein binding
          ATPase binding
          Lys63-specific deubiquitinase activity
          Lys48-specific deubiquitinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-n8jtk:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0