Search Thermo Fisher Scientific
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253877.1 | NP_001240806.1 | 6 | 1063 | ||
NM_001253878.1 | NP_001240807.1 | 6 | 782 | |||
NM_001253879.1 | NP_001240808.1 | 5 | 705 | |||
NM_001253880.1 | NP_001240809.1 | 3 | 420 | |||
NM_013827.3 | NP_038855.2 | 5 | 701 | |||
GenBank mRNA | AB221622.1 | - | 2 | 108 | ||
AK030752.1 | - | 5 | 651 | |||
AK031409.1 | - | 7 | 770 | |||
AK032798.1 | - | 6 | 1024 | |||
AK032819.1 | - | 6 | 1025 | |||
AK137539.1 | - | 7 | 1010 | |||
AK139685.1 | - | 6 | 783 | |||
BC024889.1 | - | 5 | 673 | |||
BC029076.1 | - | 5 | 612 | |||
BC050826.1 | - | 5 | 655 | |||
BC057163.1 | - | 5 | 498 | |||
BC071223.1 | - | 3 | 173 | |||
BC092237.1 | - | 5 | 633 | 30637248 | ||
BC100340.1 | - | 3 | 351 | 30715034 | ||
S78454.1 | - | 2 | 108 |