Search
Search

Oat| Gene Symbol: Oat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016978.2 | NP_058674.1 | 2 | 284 | ||
| GenBank mRNA | AK128914.1 | - | 2 | 275 | ||
| AK145877.1 | - | 2 | 285 | |||
| AK146413.1 | - | 2 | 229 | |||
| AK168851.1 | - | 2 | 233 | |||
| AK185243.1 | - | 1 | 68 | |||
| AK185430.1 | - | 1 | 68 | |||
| BC008119.1 | - | 2 | 217 | 3498240 | ||
| X64837.1 | - | 1 | 258 | |||