Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GATA2 SIRNA Assays ›
Gene Symbol:
Gata2
Gene Name:
GATA binding protein 2
Gene Aliases:
Gata-2
Chromosome Location:
Chr.6: 88192578 - 88207032 on Build GRCm38
UniGene ID:
Mm.491156
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
157707
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Gata2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008090.5
NP_032116.4
6
3137
GenBank mRNA
AB000096.2
-
6
3029
AK004551.1
-
6
3134
AK154981.1
-
6
3138
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_033442, AY032734, BC061745, FQ212304.
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
urogenital system development
cell fate determination
neuron migration
embryonic placenta development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
phagocytosis
positive regulation of cytosolic calcium ion concentration
ventral spinal cord interneuron differentiation
cell differentiation in hindbrain
commitment of neuronal cell to specific neuron type in forebrain
central nervous system neuron development
pituitary gland development
hemopoiesis
cell differentiation
neuron differentiation
somatic stem cell population maintenance
regulation of histone acetylation
eosinophil fate commitment
inner ear morphogenesis
mast cell degranulation
negative regulation of fat cell differentiation
negative regulation of myeloid cell differentiation
positive regulation of erythrocyte differentiation
negative regulation of macrophage differentiation
positive regulation of megakaryocyte differentiation
positive regulation of neuron differentiation
negative regulation of Notch signaling pathway
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
cell maturation
neuron fate commitment
homeostasis of number of cells within a tissue
positive regulation of phagocytosis
positive regulation of phagocytosis, engulfment
definitive hemopoiesis
semicircular canal development
negative regulation of fat cell proliferation
GABAergic neuron differentiation
negative regulation of neural precursor cell proliferation
regulation of forebrain neuron differentiation
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
enhancer sequence-specific DNA binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
zinc ion binding
sequence-specific DNA binding
metal ion binding
C2H2 zinc finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0