Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRIN2A SIRNA Assays ›
Gene Symbol:
Grin2a
Gene Name:
glutamate receptor, ionotropic, NMDA2A (epsilon 1)
Gene Aliases:
GluN2A, NMDAR2A, NR2A
Chromosome Location:
Chr.16: 9573737 - 9996500 on Build GRCm38
UniGene ID:
Mm.2953
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
157897
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Grin2a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008170.2
NP_032196.2
8
1885
GenBank mRNA
D10217.2
-
8
1885
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
action potential
startle response
response to amphetamine
transport
ion transport
cation transport
calcium ion transport
chemical synaptic transmission
learning or memory
learning
memory
locomotory behavior
protein localization
visual learning
response to wounding
positive regulation of cell death
sensory perception of pain
neurogenesis
sleep
directional locomotion
ion transmembrane transport
regulation of ion transmembrane transport
ionotropic glutamate receptor signaling pathway
synaptic transmission, glutamatergic
locomotion
negative regulation of protein catabolic process
regulation of membrane potential
dopamine metabolic process
serotonin metabolic process
response to drug
positive regulation of apoptotic process
response to ethanol
regulation of synaptic plasticity
regulation of long-term neuronal synaptic plasticity
rhythmic process
modulation of synaptic transmission
detection of mechanical stimulus involved in sensory perception of pain
protein tetramerization
regulation of sensory perception of pain
regulation of postsynaptic membrane potential
excitatory postsynaptic potential
long-term synaptic potentiation
calcium ion transmembrane transport
cellular response to amino acid stimulus
cellular response to zinc ion
cation transmembrane transport
positive regulation of excitatory postsynaptic potential
receptor activity
ionotropic glutamate receptor activity
NMDA glutamate receptor activity
receptor binding
ion channel activity
extracellular-glutamate-gated ion channel activity
cation channel activity
calcium channel activity
protein binding
zinc ion binding
glutamate binding
protein kinase binding
voltage-gated cation channel activity
protein complex binding
glutamate receptor binding
neurotransmitter binding
metal ion binding
protein heterodimerization activity
protein dimerization activity
cell adhesion molecule binding
ATPase binding
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0