Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EDN1 SIRNA Assays ›
          Gene Symbol:
          Edn1
          Gene Name:
          endothelin 1
          Gene Aliases:
          ET-1, preproET
          Chromosome Location:
          Chr.13: 42301270 - 42307989 on Build GRCm38
          UniGene ID:
          Mm.14543
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 158342

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Edn1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010104.3 NP_034234.1 4, 5 1123
          GenBank mRNA AB081657.1 - 4, 5 509
          AK040778.1 - 4, 5 781
          AK152647.1 - 4, 5 774
          AK153437.1 - 4, 5 778
          BC029547.1 - 4, 5 917 5059522
          D43775.1 - 4, 5 834
          U35233.1 - 4, 5 528

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          prostaglandin biosynthetic process
          patterning of blood vessels
          response to hypoxia
          in utero embryonic development
          histamine secretion
          regulation of systemic arterial blood pressure by endothelin
          regulation of pH
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          protein kinase C-activating G-protein coupled receptor signaling pathway
          cell-cell signaling
          heart development
          respiratory gaseous exchange
          body fluid secretion
          regulation of blood pressure
          positive regulation of cell proliferation
          dorsal/ventral pattern formation
          positive regulation of heart rate
          positive regulation of cardiac muscle hypertrophy
          positive regulation of receptor biosynthetic process
          neural crest cell development
          phosphatidylinositol 3-kinase signaling
          artery smooth muscle contraction
          vein smooth muscle contraction
          glucose transport
          cell growth
          regulation of vasoconstriction
          sensory perception of pain
          calcium-mediated signaling
          peptide hormone secretion
          nitric oxide transport
          positive regulation of cell migration
          negative regulation of cAMP biosynthetic process
          phospholipase D-activating G-protein coupled receptor signaling pathway
          negative regulation of cellular protein metabolic process
          positive regulation of prostaglandin secretion
          negative regulation of smooth muscle cell apoptotic process
          intracellular signal transduction
          positive regulation of urine volume
          positive regulation of renal sodium excretion
          epithelial fluid transport
          vasoconstriction
          protein kinase C deactivation
          middle ear morphogenesis
          positive regulation of odontogenesis
          superoxide anion generation
          rhythmic excitation
          positive regulation of MAP kinase activity
          positive regulation of JUN kinase activity
          positive regulation of cell size
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of smooth muscle contraction
          positive regulation of hormone secretion
          negative regulation of hormone secretion
          inositol phosphate-mediated signaling
          blood vessel morphogenesis
          positive regulation of smooth muscle cell proliferation
          regulation of blood vessel size
          cartilage development
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          negative regulation of nitric-oxide synthase biosynthetic process
          membrane depolarization
          regulation of sensory perception of pain
          positive regulation of sarcomere organization
          positive regulation of prostaglandin-endoperoxide synthase activity
          positive regulation of neutrophil chemotaxis
          receptor binding
          cytokine activity
          hormone activity
          protein binding
          endothelin A receptor binding
          endothelin B receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0