Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gbp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010259.2 | NP_034389.2 | 6 | 905 | ||
GenBank mRNA | AK077796.1 | - | 6 | 906 | ||
BC108990.2 | - | 950 | 40047769 | |||
EF494422.1 | - | 6 | 905 | |||
M55544.1 | - | 889 | ||||
M63961.1 | - | 983 |