Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173533.1 | NP_001167004.1 | 4, 5 | 758 | ||
NM_001173534.1 | NP_001167005.1 | 4, 5 | 749 | |||
NM_001184781.1 | NP_001171710.1 | 5, 6 | 872 | |||
NM_005137.2 | NP_005128.1 | 5, 6 | 881 | |||
NR_033674.1 | 4, 5 | 766 | ||||
GenBank mRNA | AK291670.1 | - | 5, 6 | 839 | ||
AK295697.1 | - | 4, 5 | 724 | |||
AK296337.1 | - | 4, 5 | 707 | |||
AK303130.1 | - | 5, 6 | 791 | |||
AK304382.1 | - | 4, 5 | 716 | |||
BC020339.1 | - | 5, 6 | 828 | |||
BC032430.1 | - | 5, 6 | 817 | 5243412 | ||
BC035425.1 | - | 5, 6 | 828 | |||
BC040500.1 | - | 5, 6 | 876 | 5272271 | ||
CR456433.1 | - | 5, 6 | 653 | |||
D79985.1 | - | 5, 6 | 818 | |||
FJ032369.1 | - | 5, 6 | 633 | |||
L77560.1 | - | 5, 6 | 782 | |||
X83545.1 | - | 5, 6 | 782 | |||
X84076.1 | - | 5, 6 | 779 |