Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Grb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010345.4 | NP_034475.2 | 5 | 772 | ||
GenBank mRNA | AF022072.1 | - | 5 | 746 | ||
AK030727.1 | - | 5 | 707 | |||
AK032927.1 | - | 5 | 753 | |||
AK080969.1 | - | 1 | 1206 | |||
AK163841.1 | - | 5 | 753 | |||
BC053842.1 | - | 5 | 739 |