Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DMTN SIRNA Assays ›
          Gene Symbol:
          Dmtn
          Gene Name:
          dematin actin binding protein
          Gene Aliases:
          AI325486, Epb4.9, Epb49, dematin
          Chromosome Location:
          Chr.14: 70602184 - 70636048 on Build GRCm38
          UniGene ID:
          Mm.210863
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 159212

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dmtn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252662.1 NP_001239591.1 16 2301
          NM_001252663.1 NP_001239592.1 15 2235
          NM_001252664.1 NP_001239593.1 15 2128
          NM_001252665.1 NP_001239594.1 14 1961
          NM_001252666.1 NP_001239595.1 13 1994
          NM_013514.4 NP_038542.1 16 2176
          GenBank mRNA AF079846.3 - 14 1905
          AF155547.1 - 15 1971
          AK143840.1 - 16 2295
          AK149301.1 - 15 1938
          AK155927.1 - 15 2231
          AK158744.1 - 15 2059
          AK162866.1 - 14 1913
          AK165204.1 - 2294
          AK165294.1 - 15 2227
          AK165709.1 - 1 574
          BC016897.1 - 16 2176 4240747
          BC037021.1 - 13 1981 5366040

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          cytoskeleton organization
          regulation of cell shape
          regulation of lamellipodium assembly
          positive regulation of fibroblast migration
          negative regulation of peptidyl-threonine phosphorylation
          negative regulation of cell-substrate adhesion
          actin cytoskeleton organization
          positive regulation of blood coagulation
          regulation of actin cytoskeleton organization
          negative regulation of peptidyl-serine phosphorylation
          calcium-mediated signaling using intracellular calcium source
          calcium-mediated signaling using extracellular calcium source
          erythrocyte development
          negative regulation of peptidyl-tyrosine phosphorylation
          actin filament bundle assembly
          regulation of filopodium assembly
          actin filament capping
          negative regulation of focal adhesion assembly
          protein secretion by platelet
          cellular response to calcium ion
          cellular response to cAMP
          positive regulation of wound healing
          negative regulation of protein targeting to membrane
          actin filament reorganization
          negative regulation of substrate adhesion-dependent cell spreading
          positive regulation of substrate adhesion-dependent cell spreading
          positive regulation of platelet aggregation
          positive regulation of integrin-mediated signaling pathway
          actin binding
          receptor binding
          spectrin binding
          protein self-association

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-w466k:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0