Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other H2-KE6 SIRNA Assays ›
Gene Symbol:
H2-Ke6
Gene Name:
H2-K region expressed gene 6
Gene Aliases:
D17H6S112E, H-2Ke6, Hsd17b8, Ke-6, Ke6, Ring2
Chromosome Location:
Chr.17: 34026033 - 34028055 on Build GRCm38
UniGene ID:
Mm.275452
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
159272
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: H2-Ke6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013543.2
NP_038571.2
9
941
GenBank mRNA
BC086927.1
-
9
941
6775958
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_212529, XM_006255965, XM_006255964, XM_006255966, BC091424.
Panther Classification:
Molecular Function -
oxidoreductase
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
steroid biosynthetic process
estrogen biosynthetic process
metabolic process
androgen metabolic process
estrogen metabolic process
oxidation-reduction process
C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
3-hydroxyacyl-CoA dehydrogenase activity
aldo-keto reductase (NADP) activity
estradiol 17-beta-dehydrogenase activity
isocitrate dehydrogenase activity
mevaldate reductase activity
gluconate dehydrogenase activity
steroid dehydrogenase activity
oxidoreductase activity
epoxide dehydrogenase activity
5-exo-hydroxycamphor dehydrogenase activity
2-hydroxytetrahydrofuran dehydrogenase activity
acetoin dehydrogenase activity
phenylcoumaran benzylic ether reductase activity
D-xylose:NADP reductase activity
L-arabinose:NADP reductase activity
D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
steroid dehydrogenase activity, acting on the CH-CH group of donors
(R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
3-hydroxymenthone dehydrogenase activity
very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
dihydrotestosterone 17-beta-dehydrogenase activity
(R)-2-hydroxyisocaproate dehydrogenase activity
L-arabinose 1-dehydrogenase (NADP+) activity
L-xylulose reductase (NAD+) activity
testosterone dehydrogenase (NAD+) activity
3-ketoglucose-reductase activity
(R)-2-hydroxyglutarate dehydrogenase activity
D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0