Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNRNPK SIRNA Assays ›
          Gene Symbol:
          Hnrnpk
          Gene Name:
          heterogeneous nuclear ribonucleoprotein K
          Gene Aliases:
          Hnrpk, KBBP, NOVA
          Chromosome Location:
          Chr.13: 58391132 - 58403343 on Build GRCm38
          UniGene ID:
          Mm.142872
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 159505

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hnrnpk
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301340.1 NP_001288269.1 12 1213
          NM_001301341.1 NP_001288270.1 14 1340
          NM_001301343.1 NP_001288272.1 12 1199
          NM_001301344.1 NP_001288273.1 11 1141
          NM_001301345.1 NP_001288274.1 11 1141
          NM_025279.3 NP_079555.1 12 1213
          GenBank mRNA AK011428.1 - 12 1218
          AK051313.1 - 11 1135
          AK078777.1 - 14 1341
          AK088462.1 - 11 1128
          AK128891.1 - 12 1206
          AK135519.1 - 12 1127
          AK144245.1 - 8423
          AK150848.1 - 12 1193
          AK151689.1 - 11 1095
          AK152927.1 - 11 1095
          AK160615.1 - 12 1183
          AK160887.1 - 12 1215
          AK166656.1 - 12 1218
          AK166927.1 - 12 1171
          AK167600.1 - 12 1204
          AK168918.1 - 8 1116
          AK169064.1 - 12 1203
          AK205281.1 - 3 333
          AK208657.1 - 1 73
          BC006694.1 - 12 1180 3583037
          BC089328.1 - 12 1136 6402614
          EU604547.1 - 10 994

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_057141, XM_006253552, XM_006253554, XM_006253553, XM_006253555, and others.

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ovarian follicle development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          mRNA processing
          RNA splicing
          positive regulation of neuron projection development
          regulation of low-density lipoprotein particle clearance
          cellular response to insulin stimulus
          positive regulation of RNA splicing
          negative regulation of apoptotic process
          positive regulation of low-density lipoprotein particle receptor biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of mRNA splicing, via spliceosome
          positive regulation of receptor-mediated endocytosis
          positive regulation of synaptic transmission
          positive regulation of dendritic spine development
          cellular response to organic cyclic compound
          regulation of lipid transport by positive regulation of transcription from RNA polymerase II promoter
          positive regulation of synapse maturation
          positive regulation of long-term synaptic potentiation
          regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of dendrite extension
          positive regulation of protein localization to cell surface
          negative regulation of branching morphogenesis of a nerve
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          core promoter proximal region DNA binding
          nucleic acid binding
          DNA binding
          single-stranded DNA binding
          RNA binding
          mRNA 3'-UTR binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline