Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CLEC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005127.2 | NP_005118.2 | 5 | 1099 | ||
GenBank mRNA | AB015628.1 | - | 5 | 592 | ||
BC005254.1 | - | 5 | 593 | 3996700 | ||
BG540007.1 | - | 1 | 338 | |||
CB160326.1 | - | 3 | 322 | |||
X96719.1 | - | 5 | 574 |