Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007462.3 | NP_031488.2 | 14 | 1976 | ||
GenBank mRNA | AK046280.1 | - | 14 | 1966 | ||
AK080907.1 | - | 14 | 1976 | |||
BC141141.1 | - | 13 | 1689 | 9055753 | ||
M88127.1 | - | 13 | 1665 |