Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Arf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031259.1 | - | 5 | 757 | ||
AK042977.1 | - | 5 | 797 | |||
AK053123.1 | - | 5 | 1215 | |||
AK077591.1 | - | 5 | 770 | |||
AK167133.1 | - | 5 | 767 | |||
AK167601.1 | - | 5 | 771 | |||
AK167653.1 | - | 5 | 770 | |||
AK169094.1 | - | 5 | 767 | |||
AK169412.1 | - | 5 | 767 | |||
AK178585.1 | - | 1 | 223 | |||
BC010487.1 | - | 5 | 1050 | 3483748 | ||
D87899.1 | - | 5 | 762 |