Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DDX5 SIRNA Assays ›
          Gene Symbol:
          Ddx5
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box polypeptide 5
          Gene Aliases:
          2600009A06Rik, G17P1, HUMP68, Hlr1, p68
          Chromosome Location:
          Chr.11: 106780355 - 106789268 on Build GRCm38
          UniGene ID:
          Mm.491096
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 160506

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ddx5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007840.3 NP_031866.2 2, 3 392
          GenBank mRNA AK044792.1 - 2, 3 392
          AK076018.1 - 2, 3 394
          AK088887.1 - 2, 3 393
          AK129022.1 - 2, 3 392
          AK132779.1 - 2, 3 439
          AK135173.1 - 2, 3 393
          AK166045.1 - 2, 3 393
          AK166698.1 - 3, 4 525
          BC046554.1 - 2, 3 370
          BC062916.1 - 2, 3 377 6827275
          BC086320.1 - 2, 3 315
          BC108369.1 - 2, 3 237 6312791
          BC129873.1 - 2, 3 270 40110366
          BC129874.1 - 2, 3 273 40110367
          X65627.1 - 2, 3 374

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001007613, XM_008768312, XM_008768313, BC079036, FJ168767.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of alternative mRNA splicing, via spliceosome
          in utero embryonic development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          circadian rhythm
          RNA splicing
          mRNA transcription
          RNA secondary structure unwinding
          positive regulation of intracellular estrogen receptor signaling pathway
          positive regulation of DNA damage response, signal transduction by p53 class mediator
          regulation of viral genome replication
          regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          regulation of androgen receptor signaling pathway
          intrinsic apoptotic signaling pathway by p53 class mediator
          regulation of skeletal muscle cell differentiation
          nucleotide binding
          nucleic acid binding
          transcription cofactor activity
          transcription coactivator activity
          RNA binding
          RNA helicase activity
          ATP-dependent RNA helicase activity
          helicase activity
          protein binding
          calmodulin binding
          ATP binding
          hydrolase activity
          hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
          enzyme binding
          estrogen receptor binding
          pre-mRNA binding
          poly(A) RNA binding
          calcium-dependent protein binding
          androgen receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0