Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Epyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007884.2 | NP_031910.1 | 4, 5 | 468 | ||
GenBank mRNA | AK005684.1 | - | 4, 5 | 469 | ||
AK015223.1 | - | 4, 5 | 468 | |||
AK137179.1 | - | 3, 4 | 393 | |||
AK157900.1 | - | 719 | ||||
AK157901.1 | - | 3, 4 | 730 | |||
BC012695.1 | - | 3, 4 | 440 | 4037028 | ||
D78274.1 | - | 3, 4 | 383 |