Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cx3cr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009987.4 | NP_034117.3 | 2 | 735 | ||
GenBank mRNA | AF102269.1 | - | 2 | 729 | ||
AK035902.1 | - | 2 | 710 | |||
AK044557.1 | - | 2 | 736 | |||
AK045634.1 | - | 2 | 723 | |||
AK137388.1 | - | 2 | 723 | |||
AK140010.1 | - | 2 | 709 | |||
BC012653.1 | - | 2 | 691 | 4017224 |
Set Membership: |
G-protein coupled receptor |