Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dlx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010054.2 | NP_034184.1 | 1 | 570 | ||
GenBank mRNA | AK144652.1 | - | 1 | 571 | ||
BC094317.1 | - | 1 | 270 | 5718422 | ||
M80540.1 | - | 1 | 508 | |||
M83184.1 | - | 1 | 133 |