Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CREM SIRNA Assays ›
          Gene Symbol:
          Crem
          Gene Name:
          cAMP responsive element modulator
          Gene Aliases:
          ICER, ICERI
          Chromosome Location:
          Chr.18: 3266354 - 3340355 on Build GRCm38
          UniGene ID:
          Mm.5244
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 161686

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Crem
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110850.2 NP_001104320.1 5 498
          NM_001110853.2 NP_001104323.1 7 761
          NM_001110854.1 NP_001104324.2 3 140
          NM_001110855.1 NP_001104325.2 2 104
          NM_001110856.2 NP_001104326.1 7 681
          NM_001110857.2 NP_001104327.1 6 534
          NM_001110858.2 NP_001104328.1 5 498
          NM_001110859.2 NP_001104329.1 8 870
          NM_001271503.1 NP_001258432.1 6 645
          NM_001271504.1 NP_001258433.1 6 645
          NM_001271505.1 NP_001258434.1 7 723
          NM_001271506.1 NP_001258435.1 7 834
          NM_013498.3 NP_038526.2 6 534
          GenBank mRNA AJ292222.1 - 2 69
          AJ311667.1 - 3 105
          AK076797.1 - 7 684
          AY738715.1 - 5 561
          AY738716.1 - 4 414
          AY738717.1 - 5 561
          AY738718.1 - 6 597
          AY738719.1 - 6 639
          AY738720.1 - 6 657
          AY738721.1 - 6 750
          BE689077.1 - 2 104
          M60285.1 - 6 462
          S64590.1 - 6 579
          S67786.1 - 2 91

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001271102, NM_017334, NM_001110860, NM_001271248, NM_001271245, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          glycosphingolipid metabolic process
          spermatogenesis
          circadian regulation of gene expression
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          response to cAMP
          core promoter sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0