Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABCA1 SIRNA Assays ›
Gene Symbol:
Abca1
Gene Name:
ATP-binding cassette, sub-family A (ABC1), member 1
Gene Aliases:
ABC-1, Abc1
Chromosome Location:
Chr.4: 53030789 - 53159988 on Build GRCm38
UniGene ID:
Mm.277376
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
162500
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Abca1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013454.3
NP_038482.3
7, 8
1039
GenBank mRNA
AK051920.1
-
7, 8
1040
X75926.1
-
6, 7
802
Back To Top
More Information
Panther Classification:
Molecular Function -
ATP-binding cassette (ABC) transporter
primary active transporter
transporter
Gene Ontology Categories:
Function(s)
Process(es)
peptide secretion
protein lipidation
transport
phagocytosis, engulfment
lysosome organization
G-protein coupled receptor signaling pathway
cholesterol metabolic process
positive regulation of cholesterol efflux
phospholipid transport
endosomal transport
cholesterol transport
positive regulation of cAMP biosynthetic process
intracellular cholesterol transport
regulation of Cdc42 protein signal transduction
cholesterol efflux
phospholipid efflux
high-density lipoprotein particle assembly
apolipoprotein A-I-mediated signaling pathway
lipoprotein metabolic process
lipoprotein biosynthetic process
cholesterol homeostasis
reverse cholesterol transport
phospholipid translocation
interleukin-1 beta secretion
transmembrane transport
phospholipid homeostasis
response to high density lipoprotein particle
platelet dense granule organization
cellular response to lipopolysaccharide
cellular response to retinoic acid
anion transmembrane transport
nucleotide binding
receptor binding
transporter activity
protein binding
ATP binding
phospholipid transporter activity
anion transmembrane transporter activity
ATPase activity
cholesterol transporter activity
syntaxin binding
small GTPase binding
apolipoprotein binding
apolipoprotein A-I binding
apolipoprotein A-I receptor activity
ATPase activity, coupled to transmembrane movement of substances
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0