Search
Search

Ak2| Gene Symbol: Ak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033966.4 | NP_001029138.1 | 2 | 317 | ||
| NM_016895.4 | NP_058591.2 | 2 | 317 | |||
| GenBank mRNA | AB020202.1 | - | 2 | 239 | ||
| AK010936.1 | - | 2 | 236 | |||
| AK010951.1 | - | 2 | 238 | |||
| AK050133.1 | - | 2 | 278 | |||
| AK077519.1 | - | 2 | 240 | |||
| AK166976.1 | - | 2 | 228 | |||
| AK167797.1 | - | 2 | 277 | |||
| AK168056.1 | - | 2 | 240 | |||
| AK168148.1 | - | 2 | 267 | |||
| BC008610.1 | - | 2 | 227 | 3584786 | ||