Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLCG1 SIRNA Assays ›
Gene Symbol:
Plcg1
Gene Name:
phospholipase C, gamma 1
Gene Aliases:
AI894140, Cded, Plc-1, Plc-gamma1, Plcg-1
Chromosome Location:
Chr.2: 160731310 - 160775760 on Build GRCm38
UniGene ID:
Mm.44463
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
162703
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Plcg1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021280.3
NP_067255.2
2
240
GenBank mRNA
AF027185.1
-
1
141
BC057161.1
-
2
250
BC065091.1
-
2
240
6854923
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_182811, NM_002660, BC136466, BX415739, BC144136, and others.
This siRNA also targets the following transcripts in Rat: NM_013187, J03806.
Panther Classification:
Molecular Function -
phospholipase
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
lipid metabolic process
calcium ion transport
signal transduction
epidermal growth factor receptor signaling pathway
protein secretion
phospholipid catabolic process
positive regulation of epithelial cell migration
lipid catabolic process
cell migration
calcium-mediated signaling
inositol trisphosphate biosynthetic process
intracellular signal transduction
positive regulation of blood vessel endothelial cell migration
positive regulation of angiogenesis
T cell receptor signaling pathway
positive regulation of release of sequestered calcium ion into cytosol
cellular response to epidermal growth factor stimulus
regulation of store-operated calcium channel activity
phosphatidylinositol phospholipase C activity
signal transducer activity
insulin receptor binding
neurotrophin TRKA receptor binding
calcium ion binding
protein binding
phosphoric diester hydrolase activity
hydrolase activity
protein kinase binding
receptor tyrosine kinase binding
glutamate receptor binding
metal ion binding
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0